Learn More
Sino Biological Recombinant Rat TL1A/TNFSF15 Protein (hFc Tag)
Supplier:  Sino Biological 80150R01H100
Encompass_Preferred
A DNA sequence encoding the rat TNFSF15 (Q8K3Y7) (Asp94-Ile252) was expressed, fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-237-2137