Learn More
BIOHIPPO Rat TNNI1 Microsample
SDP
Catalog No. 5000315680
Encompass
Rat TNNI1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TNNI1 in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 18 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TNNI1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TNNI1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TNNI1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15680 Supplier BIOHIPPO Supplier No. BHE15212511
Rat TNNI1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TNNI1 in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 18 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TNNI1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TNNI1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TNNI1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color