Learn More
Biomatik Corporation Rat Topoisomerase II (TOP2) ELISA Kit
Supplier:  Biomatik Corporation EKU07770
Encompass_Preferred
Rat Topoisomerase II (TOP2) ELISA Kit for detecting TOP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41516. Gene ID: 360243. Target Synonyms: DNA topoisomerase 2-alpha; DNA topoisomerase II, alpha isozyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8726