Learn More
Alpco Ltd Rat Total GIP ELISA
SDP
Supplier:  Alpco Ltd 48GIPRTE01
Encompass_Preferred
Rat Total GIP ELISA is for the quantitative determination of total glucose-dependent insulinotropic polypeptide in rat EDTA plasma and culture medium supernatant. Test kit can be used for measurement of both GIP (1-42) and GIP (3-42). Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2329182