Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat Total MMP-9 DuoSet ELISA

Catalog No. DY817405
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY817405 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY817405 Supplier R&D Systems™ Supplier No. DY817405
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant rat total MMP-9.

The Rat Total MMP-9 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies rat MMP-9.

Specifications

Assay Range 78.10 to 5000 pg/mL
Conjugate Biotin
Immunoassay Kit Format 96-well Strip Plate
Product Type Solid Phase Sandwich ELISA
Sufficient For Five 96-well plates
For Use With (Application) ELISA
Gene ID (Entrez) 4318
Gene Symbol Mmp9
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase
Target MMP-9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.