Learn More
BIOHIPPO Rat tPA Microsample
SDP
Catalog No. 5000315317
Encompass
Rat tPA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of tPA in plasma It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 126 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat tPA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat tPA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat tPA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15317 Supplier BIOHIPPO Supplier No. BHE15211979
Rat tPA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of tPA in plasma It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 126 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat tPA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat tPA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat tPA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color