Learn More
Biomatik Corporation Rat Transcription Factor P65 (RELA) ELISA Kit
Supplier:  Biomatik Corporation EKU08193
Encompass_Preferred
Rat Transcription Factor P65 (RELA) ELISA Kit for detecting RELA in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7TQN4. Gene ID: 309165. Target Synonyms: Nuclear factor NF-kappa-B p65 subunit; NFKBP65; RELA; p65; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4877