Learn More
Immunology Consultants Rat Transferrin ELISA Kit
SDP
Supplier:  Immunology Consultants E25TX
Encompass_Preferred
The Rat Transferrin (TX) test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring TX in rat biological samples.
Catalog No. 50-240-6555