Learn More
Biomatik Corporation Rat Transferrin Receptor 2 (TFR2) ELISA Kit
Supplier:  Biomatik Corporation EKU07793
Encompass_Preferred
Rat Transferrin Receptor 2 (TFR2) ELISA Kit for detecting TFR2 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B2GUY2. Gene ID: 288562. Target Synonyms: HFE3; TFRC2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6655