Learn More
Biomatik Corporation Rat Transferrin (TF) CLIA Kit
Supplier:  Biomatik Corporation EKU08953
Encompass_Preferred
Rat Transferrin (TF) CLIA Kit for detecting TF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 54.9-40,000pg/ml. Sensitivity: 20.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P12346. Gene ID: 24825. Target Synonyms: TRF; Siderophilin; Serotransferrin; Beta-1 metal-binding globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6291