Learn More
Biomatik Corporation Rat Transforming Growth Factor Beta 3 (TGFb3) CLIA Kit
Supplier:  Biomatik Corporation EKU09709
Encompass_Preferred
Rat Transforming Growth Factor Beta 3 (TGFb3) CLIA Kit for detecting TGFb3 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.62pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q07258. Gene ID: 25717. Target Synonyms: TGF-B3; LAP; Latency-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8459