Learn More
American Research Products Inc Rat Transforming Growth Factor
Catalog No. 5000326307
Encompass
Rat Transforming Growth Factor b1 (TGF-b1) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26307 Supplier American Research Products Inc Supplier No. CSBE04727R
Rat Transforming Growth Factor b1 (TGF-b1) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich