Learn More
American Research Products Inc Rat transforming growth factor
Catalog No. 5000326461
Encompass
Rat transforming growth factor a TGF-a ELISA Kit from CUSABIO Detection Range 7 8 pg/ml-500pg/ml Sensitivity 1 95 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26461 Supplier American Research Products Inc Supplier No. CSBE07289R
Rat transforming growth factor a TGF-a ELISA Kit from CUSABIO Detection Range 7 8 pg/ml-500pg/ml Sensitivity 1 95 pg/ml Sample serum plasma tissue homogenates Method Sandwich