Learn More
American Research Products Inc Rat Transforming Growth factor
Catalog No. 5000329775
Encompass
Rat Transforming Growth factor b2 TGF-b2 ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29775 Supplier American Research Products Inc Supplier No. CSBE12111R
Rat Transforming Growth factor b2 TGF-b2 ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich