Learn More
Biomatik Corporation Rat Transient Receptor Potential Cation Channel Subfamily M, Member 4 (TRPM4) ELISA Kit
Supplier:  Biomatik Corporation EKU07840
Encompass_Preferred
Rat Transient Receptor Potential Cation Channel Subfamily M, Member 4 (TRPM4) ELISA Kit for detecting TRPM4 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ESQ5. Gene ID: 171143. Target Synonyms: TRPM4B; Melastatin-4; Calcium-activated non-selective cation channel 1; Long transient receptor potential channel 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5865