Learn More
Biomatik Corporation Rat Transient Receptor Potential Cation Channel Subfamily A, Member 1 (TRPA1) ELISA Kit
Supplier:  Biomatik Corporation EKU12413
Encompass_Preferred
Rat Transient Receptor Potential Cation Channel Subfamily A, Member 1 (TRPA1) ELISA Kit for detecting TRPA1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6RI86. Gene ID: 312896. Target Synonyms: ANKTM1; Ankyrin-like with transmembrane domains protein 1; Transformation-sensitive protein p120. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6050