Learn More
Biomatik Corporation Rat Transketolase (TKT) ELISA Kit
Supplier:  Biomatik Corporation EKU07849
Encompass_Preferred
Rat Transketolase (TKT) ELISA Kit for detecting TKT in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50137. Gene ID: 64524. Target Synonyms: TK; Wernicke-Korsakoff Syndrome. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8720