Learn More
American Research Products Inc Rat Translationally controlled
Catalog No. 5000328193
Encompass
Rat Translationally-controlled tumor protein (TPT1) ELISA kit from CUSABIO Detection Range 39 pg/mL-2500 pg/mL Sensitivity 9 75 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28193 Supplier American Research Products Inc Supplier No. CSBEL024134RA
Rat Translationally-controlled tumor protein (TPT1) ELISA kit from CUSABIO Detection Range 39 pg/mL-2500 pg/mL Sensitivity 9 75 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich