Learn More
Biomatik Corporation Rat Transmembrane Protein 27 (TMEM27) ELISA Kit
Supplier:  Biomatik Corporation EKU07862
Encompass_Preferred
Rat Transmembrane Protein 27 (TMEM27) ELISA Kit for detecting TMEM27 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ESG3. Gene ID: 57395. Target Synonyms: NX17; Collectrin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4942