Learn More
World Precision Instrument Rat Transverse Clamps, Set of 4
SDP
Catalog No. 502539134
Encompass
The design was inspired by the need for a practical, versatile, and economical instrument for stabilizing the spinal column. The Spinal Adaptors are accessories to be mounted on the 505211, 505351 or other brands of stereotaxic instruments, converting them for precise spinal surgery applications.
Catalog No. 50-253-9134 Supplier World Precision Instrument Supplier No. 505291
The design was inspired by the need for a practical, versatile, and economical instrument for stabilizing the spinal column. The Spinal Adaptors are accessories to be mounted on the 505211, 505351 or other brands of stereotaxic instruments, converting them for precise spinal surgery applications.