Learn More
Biomatik Corporation Rat Trefoil Factor 1 (TFF1) ELISA Kit
Supplier:  Biomatik Corporation EKU07870
Encompass_Preferred
Rat Trefoil Factor 1 (TFF1) ELISA Kit for detecting TFF1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.12-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62737. Gene ID: 79129. Target Synonyms: BCEI; HP1.A; HPS2; PS2; PNR2; Breast Cancer,Estrogen-Inducible Sequence Expressed In; Breast cancer estrogen-inducible protein; Polypeptide P1.A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6649