Learn More
American Research Products Inc Rat triggering receptor eXpres
Catalog No. 5000328267
Encompass
Rat triggering receptor eXpressed on myeloid cells 2 (TREM2) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28267 Supplier American Research Products Inc Supplier No. CSBEL024405RA
Rat triggering receptor eXpressed on myeloid cells 2 (TREM2) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich