Learn More
MyBioSource Rat Triiodothyronine (T3) ELISA Kit
SDP
Catalog No. NC2396113
Encompass_Preferred
The MyBioSource Rat Triiodothyronine (T3) ELISA Kit quantitatively measures T3 in various biological fluids using a sandwich enzyme immunoassay. A 96-well plate coated with a T3-specific antibody captures T3 from samples. Post-wash, biotinylated anti-T3 antibody and HRP-conjugated streptavidin are added. TMB substrate induces a color change proportional to T3 levels, measured at 450 nm after adding the Stop Solution.
Catalog No. NC2396113 Supplier MyBioSource Supplier No. MBS26128596STRIPWELLS
The MyBioSource Rat Triiodothyronine (T3) ELISA Kit quantitatively measures T3 in various biological fluids using a sandwich enzyme immunoassay. A 96-well plate coated with a T3-specific antibody captures T3 from samples. Post-wash, biotinylated anti-T3 antibody and HRP-conjugated streptavidin are added. TMB substrate induces a color change proportional to T3 levels, measured at 450 nm after adding the Stop Solution.