Learn More
American Research Products Inc Rat Tripartite motif containin
Catalog No. 5000327544
Encompass
Rat Tripartite motif-containing protein 72 (TRIM72) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 7 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27544 Supplier American Research Products Inc Supplier No. CSBEL024511RA
Rat Tripartite motif-containing protein 72 (TRIM72) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 7 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich