Learn More
Alpco Ltd Rat TSH ELISA
SDP
Supplier:  Alpco Ltd 55TSHRTE01
Encompass_Preferred
The Rat TSH ELISA is an enzyme immunoassay for the quantitative measurement of TSH in rat serum.
  • Includes standards
  • No cross reaction with Rat LH and FSH
  • User friendly test prodecure
  • See Rat Control Set 55-DEV99RC for controls
For research use only. Not for use in diagnostic procedures.
Catalog No. NC2318924