Learn More
BIOHIPPO Rat TSHR Microsample
SDP
Catalog No. 5000314460
Encompass
Rat TSHR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TSHR in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 115 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TSHR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TSHR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TSHR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14460 Supplier BIOHIPPO Supplier No. BHE15210980
Rat TSHR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TSHR in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 115 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TSHR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TSHR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TSHR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color