Learn More
BIOHIPPO Rat TSP-1 Microsample
SDP
Catalog No. 5000314776
Encompass
Rat TSP-1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TSP-1 in Plasma It provides a detection range of 1 56-100 ng/mL with a sensitivity of 0 47 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TSP-1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TSP-1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TSP-1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14776 Supplier BIOHIPPO Supplier No. BHE15212645
Rat TSP-1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TSP-1 in Plasma It provides a detection range of 1 56-100 ng/mL with a sensitivity of 0 47 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TSP-1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TSP-1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TSP-1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color