Learn More
Sigma Aldrich Fine Chemicals Biosciences Rat Tumor Necrosis factor
Catalog No. 502906599
Encompass
The Rat TNf-alpha ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat TNf-alpha cell lysate and tissue lysate.
Catalog No. 50-290-6599 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. RAB04801KT
The Rat TNf-alpha ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat TNf-alpha cell lysate and tissue lysate.