Learn More
Cloud Clone Corp Rat Tumor Necrosis Factor Alpha (TNFa) (Sandwich ELISA) ELISA Kit
Catalog No. NC4011103
Encompass_Preferred
Rat Tumor Necrosis Factor Alpha (TNFa) (Sandwich ELISA) ELISA Kit measures Rat TNFa in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Tumor Necrosis Factor Alpha (TNFa).No significant cross-reactivity or interference between Tumor Necrosis Factor Alpha (TNFa) and analogues was observed.Detection Range: 15.6-1,000pg/mLSensitivity:The minimum detectable dose of this kit is typically less than 5.9pg/mLDetection menthod: ColormetricGene ID:24835
Catalog No. NC4011103 Supplier Cloud Clone Corp Supplier No. SEA133RA
Rat Tumor Necrosis Factor Alpha (TNFa) (Sandwich ELISA) ELISA Kit measures Rat TNFa in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Tumor Necrosis Factor Alpha (TNFa).No significant cross-reactivity or interference between Tumor Necrosis Factor Alpha (TNFa) and analogues was observed.Detection Range: 15.6-1,000pg/mLSensitivity:The minimum detectable dose of this kit is typically less than 5.9pg/mLDetection menthod: ColormetricGene ID:24835