Learn More
American Research Products Inc Rat tumor necrosis factor indu
Catalog No. 5000327003
Encompass
Rat tumor necrosis factor-inducible gene 6 protein (TSG-6) elisa kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27003 Supplier American Research Products Inc Supplier No. CSBE17373R
Rat tumor necrosis factor-inducible gene 6 protein (TSG-6) elisa kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich