Learn More
American Research Products Inc Rat tumor necrosis factor lig
Catalog No. 5000327815
Encompass
Rat tumor necrosis factor (ligand) superfamily member 14 (TNFSF14) ELISA kit from CUSABIO Detection Range 15 6 pg/ml - 1000pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27815 Supplier American Research Products Inc Supplier No. CSBEL023991RA
Add to Cart
Add to Cart
Rat tumor necrosis factor (ligand) superfamily member 14 (TNFSF14) ELISA kit from CUSABIO Detection Range 15 6 pg/ml - 1000pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich