Learn More
American Research Products Inc Rat Tumor necrosis factor liga
Catalog No. 5000327901
Encompass
Rat Tumor necrosis factor ligand superfamily member 15 (TNFSF15) ELISA kit from CUSABIO Detection Range 25 pg/mL-1600 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27901 Supplier American Research Products Inc Supplier No. CSBEL023992RA
Rat Tumor necrosis factor ligand superfamily member 15 (TNFSF15) ELISA kit from CUSABIO Detection Range 25 pg/mL-1600 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich