Learn More
American Research Products Inc Rat Tumor necrosis factor solu
Catalog No. 5000326669
Encompass
Rat Tumor necrosis factor soluble receptor I TNFsR-I ELISA KIT from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26669 Supplier American Research Products Inc Supplier No. CSBE07381R
Rat Tumor necrosis factor soluble receptor I TNFsR-I ELISA KIT from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich