Learn More
American Research Products Inc Rat Tumor necrosis factor solu
Supplier:  American Research Products Inc CSBE07380R
Encompass
Rat Tumor necrosis factor soluble receptor II TNFsR-II ELISA KIT from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30186