Learn More
Invitrogen™ RAT TWEAK R ELISA Kit
Description
Rat TWEAK R quantitates rat TWEAK R in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat TWEAK R.
Specifications
Specifications
| Accession Number | AI255180; C87282; CD266; FGF-inducible 14; Fgfrp2; fibroblast growth factor regulated protein 2; fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; FN14; HPIP; TNF receptor superfamily member 12 A; Tnfrsf12a; Tumor necrosis factor receptor superfamily member 12 A; tumor necrosis factor receptor superfamily, member 12 A; TweakR; TWEAK-R; tweak-receptor; type I transmembrane protein Fn14 |
| Assay Range | 0.3-80 ng/mL |
| Assay Sensitivity | 0.3 ng/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Rat |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | AI255180; C87282; CD266; FGF-inducible 14; Fgfrp2; fibroblast growth factor regulated protein 2; fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; FN14; HPIP; TNF receptor superfamily member 12A; TNFRSF12A; Tumor necrosis factor receptor superfamily member 12A; tumor necrosis factor receptor superfamily, member 12A; TweakR; TWEAK-R; Tweak-receptor; type I transmembrane protein Fn14AI255180; C87282; CD266; FGF-inducible 14; Fgfrp2; fibroblast growth factor regulated protein 2; fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; FN14; HPIP; TNF receptor superfamily member 12A; TNFRSF12A; Tumor necrosis factor receptor superfamily member 12A; tumor necrosis factor receptor superfamily, member 12A; TweakR; TWEAK-R; Tweak-receptor; type I transmembrane protein Fn14; |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.