Learn More
Biomatik Corporation Rat Tyrosinase (TYR) ELISA Kit
Supplier:  Biomatik Corporation EKU07998
Encompass_Preferred
Rat Tyrosinase (TYR) ELISA Kit for detecting TYR in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D4A9G4. Gene ID: 308800. Target Synonyms: OCAIA; OCA1A; OCA1; CMM8; Oculocutaneous Albinism IA; Monophenol monooxygenase; Tumor rejection antigen AB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7203