Learn More
Biotang Inc Rat ultrasensitive thyroid-stimulating hormone,U-TSH ELISA Kit, 96T
Catalog No. 50155455 Shop All BIOTANG Inc Products
Encompass_Preferred
Rat ultrasensitive thyroid-stimulating hormone,U-TSH ELISA Kit, 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.
Catalog No. 50-155-455 Supplier Biotang Inc Supplier No. R6762
Rat ultrasensitive thyroid-stimulating hormone,U-TSH ELISA Kit, 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.