Learn More
American Research Products Inc Rat Ubiquitin carboXyl termina
Catalog No. 5000327587
Encompass
Rat Ubiquitin carboXyl-terminal hydrolase isozyme L1 (UCHL1) ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27587 Supplier American Research Products Inc Supplier No. CSBEL025541RA
Rat Ubiquitin carboXyl-terminal hydrolase isozyme L1 (UCHL1) ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich