Learn More
Biomatik Corporation Rat UDP Glucuronosyltransferase 2 Family, Polypeptide B7 (UGT2B7) ELISA Kit
Catalog No. 502415536
Encompass_Preferred
Rat UDP Glucuronosyltransferase 2 Family, Polypeptide B7 (UGT2B7) ELISA Kit for detecting UGT2B7 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62789. Gene ID: 286989. Target Synonyms: UGTB2B9; UDPGTh-2; 3,4-catechol estrogen-specific UDPGT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5536 Supplier Biomatik Corporation Supplier No. EKU08032
Rat UDP Glucuronosyltransferase 2 Family, Polypeptide B7 (UGT2B7) ELISA Kit for detecting UGT2B7 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62789. Gene ID: 286989. Target Synonyms: UGTB2B9; UDPGTh-2; 3,4-catechol estrogen-specific UDPGT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.