Learn More
World Precision Instrument Rat Ultra Precise Dual Stereotaxic Instrument
SDP
Catalog No. 502539195
Encompass
Classic and Proven U-Frame Design for Small Animals
The WPI Rat Stereotaxic Instruments has been a standard in research labs for nearly five decades. The time-proven 'U'-Frame design provides stability and adaptability while the instrument's exclusive, triple-lead screws allow for smooth, consistent movements with rapid positioning. The WPI Rat Stereotaxic Instruments includes a 100 micron 3-axes, left-hand manipulator arm, rat adaptor, traditional (18˚) ear bars, and corner clamp probe holder. Available as dual, ultra-precise, digital or motorized.
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
Catalog No. 50-253-9195 Supplier World Precision Instrument Supplier No. 505217
Add to Cart
Add to Cart
Request a Quote
Classic and Proven U-Frame Design for Small Animals
The WPI Rat Stereotaxic Instruments has been a standard in research labs for nearly five decades. The time-proven 'U'-Frame design provides stability and adaptability while the instrument's exclusive, triple-lead screws allow for smooth, consistent movements with rapid positioning. The WPI Rat Stereotaxic Instruments includes a 100 micron 3-axes, left-hand manipulator arm, rat adaptor, traditional (18˚) ear bars, and corner clamp probe holder. Available as dual, ultra-precise, digital or motorized.
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.