Learn More
Biomatik Corporation Rat Uncoupling Protein 3, Mitochondrial (UCP3) ELISA Kit
Supplier:  Biomatik Corporation EKU12236
Encompass_Preferred
Rat Uncoupling Protein 3, Mitochondrial (UCP3) ELISA Kit for detecting UCP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56499. Gene ID: 25708. Target Synonyms: SLC25A9; Mitochondrial,Proton Carrier; Solute carrier family 25 member 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4752