Learn More
American Research Products Inc Rat UndercarboXylated Osteocal
Catalog No. 5000328464
Encompass
Rat UndercarboXylated Osteocalcin (ucOC) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28464 Supplier American Research Products Inc Supplier No. CSBE17798R
Rat UndercarboXylated Osteocalcin (ucOC) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich