Learn More
Crystal Chem Inc Rat Urinary (NAG) Assay Kit, 96-tests
SDP
Catalog No. 501947702
Encompass_Preferred
The Rat Urinary (NAG) Assay is used to quantify levels of NAG in rat urine using only a 10 µL sample volume. Highlights: Kit uses sample size of only 10 µL, ideal for rat applications. Excellent correlation to ELISA method. Less than 15 minute procedure. Can be run manually or fully automated on a wide range of instruments. Designed for rat samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7702 Supplier Crystal Chem Inc Supplier No. 80390
The Rat Urinary (NAG) Assay is used to quantify levels of NAG in rat urine using only a 10 µL sample volume. Highlights: Kit uses sample size of only 10 µL, ideal for rat applications. Excellent correlation to ELISA method. Less than 15 minute procedure. Can be run manually or fully automated on a wide range of instruments. Designed for rat samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.