Learn More
American Research Products Inc Rat urokinase plasminogen acti
Catalog No. 5000326472
Encompass
Rat urokinase plasminogen actiVator uPA ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26472 Supplier American Research Products Inc Supplier No. CSBE07368R
Rat urokinase plasminogen actiVator uPA ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich