Learn More
Biomatik Corporation Rat Uroporphyrinogen Decarboxylase (UROD) ELISA Kit
Supplier:  Biomatik Corporation EKU08060
Encompass_Preferred
Rat Uroporphyrinogen Decarboxylase (UROD) ELISA Kit for detecting UROD in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32362. Target Synonyms: PCT; Uroporphyrinogen III Decarboxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5722