Learn More
Instech Rat 1ch magnetic VAB tether kit: VABR1T/22, 375/22PS, 24in CoEx, LS22
SDP
Supplier:  Instech KVABR1T/22
Encompass
For tethered and intermittent access.
This transcutaneous button features an external port with a septum for quick, clean and repeated access to a rat's implanted catheter. Applications include bolus or continuous dosing, blood sampling and IV self-administration. For intermittent access use a syringe fitted with a needlefree PinPort™ injector; for continuous access from outside the cage snap in a mating tether.
The design uses magnets for low stress connection, and smaller PinPorts™ so that tethers, injectors and caps are compatible with the two-channel buttons.
They are available with 22ga connectors to mate with 3Fr polyurethane rat catheters, 25ga for 2Fr catheters, or 27ga for intrathecal catheters.
Group house animals by using the protective aluminum cap — an animal welfare benefit that has never before been available to rats with externalized catheters.
NOTE: Not MRI-compatible. For MRI-compatible button: VAB95BS-MRI.
Catalog No. NC2118631