Learn More
Instech Rat Vascular Access Button tether kit for ABS: 10.5in VABR1TR/25 (red port of 2ch VAB), 375/25PS, 9in 015 silicone
SDP
Catalog No. NC1322391
Encompass
The Instech KVABR1TR/25-ABS rat tether kit is designed for sampling from the red port of a two-channel VAB™ (VABR2B/22). It includes a 10.5in (27cm) red-port tether, a 25ga plastic swivel (375/25PS), 9in (23cm) of 0.015in ID silicone tubing, and 13.5in (34cm) of PU tubing. A PS95 spring (10.5in / 27cm) provides strain relief, and the 25ga connector mates exclusively with the red port. Configured for ABS2 automated blood sampling, leaving the white port unused.
Features
  • Red-port tether for 2ch rat VAB™
  • 10.5in (27cm) VABR1TR/25 tether
  • 375/25PS swivel, 25ga
  • 9in (23cm) 0.015in silicone + 13.5in (34cm) PU tubing
  • 10.5in (27cm) PS95 spring
  • ABS2™-compatible; red port only
Catalog No. NC1322391 Supplier Instech Supplier No. KVABR1TR/25ABS
The Instech KVABR1TR/25-ABS rat tether kit is designed for sampling from the red port of a two-channel VAB™ (VABR2B/22). It includes a 10.5in (27cm) red-port tether, a 25ga plastic swivel (375/25PS), 9in (23cm) of 0.015in ID silicone tubing, and 13.5in (34cm) of PU tubing. A PS95 spring (10.5in / 27cm) provides strain relief, and the 25ga connector mates exclusively with the red port. Configured for ABS2 automated blood sampling, leaving the white port unused.
Features
  • Red-port tether for 2ch rat VAB™
  • 10.5in (27cm) VABR1TR/25 tether
  • 375/25PS swivel, 25ga
  • 9in (23cm) 0.015in silicone + 13.5in (34cm) PU tubing
  • 10.5in (27cm) PS95 spring
  • ABS2™-compatible; red port only