Learn More
BIOHIPPO Rat VAP-1 Microsample
SDP
Catalog No. 5000315716
Encompass
Rat VAP-1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VAP-1 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 1 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat VAP-1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat VAP-1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat VAP-1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15716 Supplier BIOHIPPO Supplier No. BHE15210499
Rat VAP-1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VAP-1 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 1 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat VAP-1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat VAP-1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat VAP-1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color