Learn More
American Research Products Inc Rat Vascular cell adhesion pro
Catalog No. 5000330199
Encompass
Rat Vascular cell adhesion protein 1 (VCAM-1/CD106) ELISA kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30199 Supplier American Research Products Inc Supplier No. CSBE07275R
Rat Vascular cell adhesion protein 1 (VCAM-1/CD106) ELISA kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates Method Sandwich