Learn More
American Research Products Inc Rat Vascular endothelial cell
Catalog No. 5000326291
Encompass
Rat Vascular endothelial cell growth factor receptor 2 VEGFR-2/Flk-1 ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26291 Supplier American Research Products Inc Supplier No. CSBE07348R
Rat Vascular endothelial cell growth factor receptor 2 VEGFR-2/Flk-1 ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich